Assay Details
Gene Symbol: Taf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506042.2 | XP_003506090.1 | 2 - 3 | 213 | 62 | |
XM_007646497.1 | XP_007644687.1 | 2 - 3 | 213 | 62 | ||
XM_007646498.1 | XP_007644688.1 | 2 - 3 | 213 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
