Assay Details
Gene Symbol: Gnb2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644562.1 | XP_007642752.1 | 2 - 3 | 378 | 100 | |
GenBank mRNA | JP052558.1 | - | 1 - 2 | 138 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
