Assay Details
Gene Symbol: Gnb2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644562.1 | XP_007642752.1 | 5 - 6 | 731 | 80 | |
GenBank mRNA | JP052558.1 | - | 4 - 5 | 491 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
