Assay Details
| Gene Symbol: Cldn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495595.2 | XP_003495643.1 | 1 - 2 | 217 | 90 | |
| XM_007642412.1 | XP_007640602.1 | 1 - 2 | 217 | 90 | ||
More Information
Set Membership: |
|

Cldn1
Genomic Map