Assay Details
| Gene Symbol: Asf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501941.2 | XP_003501989.1 | 1 - 2 | 307 | 70 | |
| GenBank mRNA | JI863944.1 | - | 1 - 2 | 275 | 70 | |
| JP045652.1 | - | 1 - 2 | 275 | 70 | ||
More Information
Set Membership: |
|

Asf1b
Genomic Map