Assay Details
| Gene Symbol: Asf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501941.2 | XP_003501989.1 | 2 - 3 | 425 | 98 | |
| GenBank mRNA | JI863944.1 | - | 2 - 3 | 393 | 98 | |
| JP045652.1 | - | 2 - 3 | 393 | 98 | ||
More Information
Set Membership: |
|

Asf1b
Genomic Map