Assay Details
Gene Symbol: Bid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505722.2 | XP_003505770.1 | 2 - 3 | 376 | 71 | |
GenBank mRNA | JI875955.1 | - | 2 - 3 | 376 | 71 | |
JP051702.1 | - | 2 - 3 | 376 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
