Assay Details
| Gene Symbol: Tbxa2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506448.2 | XP_003506496.1 | 2 - 3 | 1070 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbxa2r| Gene Symbol: Tbxa2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506448.2 | XP_003506496.1 | 2 - 3 | 1070 | 57 | |
Set Membership: |
|