Assay Details
| Gene Symbol: Sugt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651841.1 | XP_007650031.1 | 191 | 100 | ||
| GenBank mRNA | JI877842.1 | - | 191 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sugt1| Gene Symbol: Sugt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651841.1 | XP_007650031.1 | 191 | 100 | ||
| GenBank mRNA | JI877842.1 | - | 191 | 100 | ||