Assay Details
Gene Symbol: Znf513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497055.2 | XP_003497103.1 | 2 - 3 | 230 | 80 | |
XM_007607543.1 | XP_007605733.1 | 1 - 2 | 369 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
