Assay Details
| Gene Symbol: R3hcc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497786.2 | XP_003497834.1 | 2 - 3 | 1843 | 81 | |
| XM_007617343.1 | XP_007615533.1 | 4 - 5 | 2075 | 81 | ||
| XM_007617356.1 | XP_007615546.1 | 4 - 5 | 2082 | 81 | ||
| XM_007617362.1 | XP_007615552.1 | 3 - 4 | 1929 | 81 | ||
| XM_007617371.1 | XP_007615561.1 | 4 - 5 | 2075 | 81 | ||
More Information
Set Membership: |
|

R3hcc1l
Genomic Map