Assay Details
| Gene Symbol: Polr2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506924.2 | XP_003506972.1 | 2 - 3 | 179 | 83 | |
| GenBank mRNA | JI893953.1 | - | 2 - 3 | 230 | 83 | |
| JP045904.1 | - | 2 - 3 | 230 | 83 | ||
More Information
Set Membership: |
|

Polr2k
Genomic Map