Assay Details
Gene Symbol: Mapk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495014.2 | XP_003495062.1 | 2 - 3 | 1388 | 85 | |
GenBank mRNA | JI874638.1 | - | 1 - 2 | 1181 | 85 | |
JP065360.1 | - | 1 - 2 | 1183 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
