Assay Details
Gene Symbol: LOC100762283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514013.1 | XP_003514061.1 | 4 - 5 | 515 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100762283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514013.1 | XP_003514061.1 | 4 - 5 | 515 | 74 |
Set Membership: |
![]() ![]() ![]() |