Assay Details
| Gene Symbol: Kiaa0513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654939.1 | XP_007653129.1 | 8 - 9 | 1160 | 64 | |
| XM_007654968.1 | XP_007653158.1 | 8 - 9 | 1511 | 64 | ||
| XM_007654996.1 | XP_007653186.1 | 8 - 9 | 1041 | 64 | ||
More Information
Set Membership: |
|

Kiaa0513
Genomic Map