Assay Details
Gene Symbol: Faim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497726.2 | XP_003497774.1 | 7 - 8 | 1083 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
