Assay Details
| Gene Symbol: Timm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506158.2 | XP_003506206.1 | 2 - 3 | 256 | 87 | |
| GenBank mRNA | JI870101.1 | - | 2 - 3 | 250 | 87 | |
| JP050097.1 | - | 2 - 3 | 250 | 87 | ||
More Information
Set Membership: |
|

Timm10
Genomic Map