Assay Details
Gene Symbol: Tspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508027.2 | XP_003508075.1 | 2 - 3 | 272 | 90 | |
XM_007648652.1 | XP_007646842.1 | 1 - 2 | 231 | 90 | ||
XM_007648653.1 | XP_007646843.1 | 2 - 3 | 322 | 90 | ||
XM_007648654.1 | XP_007646844.1 | 2 - 3 | 355 | 90 | ||
GenBank mRNA | JI882606.1 | - | 2 - 3 | 258 | 90 | |
JP059586.1 | - | 2 - 3 | 258 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
