Assay Details
| Gene Symbol: Bri3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509004.2 | XP_003509052.1 | 2 - 3 | 324 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bri3bp| Gene Symbol: Bri3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509004.2 | XP_003509052.1 | 2 - 3 | 324 | 59 | |
Set Membership: |
|