Assay Details
| Gene Symbol: Rp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513577.2 | XP_003513625.1 | 5 - 6 | 1233 | 84 | |
| GenBank mRNA | JI869289.1 | - | 5 - 6 | 1116 | 84 | |
| JP043609.1 | - | 5 - 6 | 1116 | 84 | ||
More Information
Set Membership: |
|

Rp2
Genomic Map