Assay Details
| Gene Symbol: Znf394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650313.1 | XP_007648503.1 | 2 - 3 | 1583 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znf394| Gene Symbol: Znf394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650313.1 | XP_007648503.1 | 2 - 3 | 1583 | 78 | |
Set Membership: |
|