Assay Details
| Gene Symbol: Ptger1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501885.2 | XP_003501933.2 | 2 - 3 | 2744 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptger1| Gene Symbol: Ptger1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501885.2 | XP_003501933.2 | 2 - 3 | 2744 | 81 | |
Set Membership: |
|