Assay Details
| Gene Symbol: Ccnd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506041.2 | XP_003506089.1 | 4 - 5 | 823 | 67 | |
| XM_007646494.1 | XP_007644684.1 | 4 - 5 | 551 | 67 | ||
| XM_007646495.1 | XP_007644685.1 | 3 - 4 | 607 | 67 | ||
| XM_007646496.1 | XP_007644686.1 | 4 - 5 | 568 | 67 | ||
| GenBank mRNA | JP051628.1 | - | 4 - 5 | 580 | 67 | |
More Information
Set Membership: |
|

Ccnd3
Genomic Map