Assay Details
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653524.1 | XP_007651714.1 | 4 - 5 | 698 | 105 | |
GenBank mRNA | JI888543.1 | - | 2 - 3 | 638 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
