Assay Details
| Gene Symbol: Hiat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653524.1 | XP_007651714.1 | 8 - 9 | 1211 | 86 | |
| GenBank mRNA | JI888543.1 | - | 6 - 7 | 1151 | 86 | |
More Information
Set Membership: |
|

Hiat1
Genomic Map