Assay Details
| Gene Symbol: Srp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643151.1 | XP_007641341.1 | 1 | 196 | 71 | |
| GenBank mRNA | JI892202.1 | - | 1 | 196 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srp19| Gene Symbol: Srp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643151.1 | XP_007641341.1 | 1 | 196 | 71 | |
| GenBank mRNA | JI892202.1 | - | 1 | 196 | 71 | |
Set Membership: |
|