Assay Details
| Gene Symbol: Tff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645006.1 | XP_007643196.1 | 2 - 3 | 90 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tff1| Gene Symbol: Tff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645006.1 | XP_007643196.1 | 2 - 3 | 90 | 60 | |
Set Membership: |
|