Assay Details
Gene Symbol: Pus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501711.2 | XP_003501759.1 | 4 - 5 | 730 | 63 | |
XM_003501712.2 | XP_003501760.1 | 4 - 5 | 517 | 63 | ||
GenBank mRNA | JI888253.1 | - | 4 - 5 | 531 | 63 | |
JP048409.1 | - | 4 - 5 | 531 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
