Assay Details
Gene Symbol: Fam196a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510496.1 | XP_003510544.1 | 3 - 4 | 1230 | 75 | |
XM_007651120.1 | XP_007649310.1 | 3 - 4 | 1230 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
