Assay Details
Gene Symbol: Slc10a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513244.2 | XP_003513292.2 | 4 - 5 | 758 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
