Assay Details
Gene Symbol: Cnih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650030.1 | XP_007648220.1 | 4 - 5 | 738 | 81 | |
XM_007650031.1 | XP_007648221.1 | 4 - 5 | 327 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
