Assay Details
| Gene Symbol: Duoxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498022.2 | XP_003498070.1 | 5 - 6 | 772 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Duoxa1| Gene Symbol: Duoxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498022.2 | XP_003498070.1 | 5 - 6 | 772 | 60 | |
Set Membership: |
|