Assay Details
Gene Symbol: Bola3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510030.2 | XP_003510078.1 | 3 - 4 | 336 | 60 | |
GenBank mRNA | JI893933.1 | - | 3 - 4 | 337 | 60 | |
JP046833.1 | - | 3 - 4 | 337 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
