Assay Details
Gene Symbol: Lxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510291.2 | XP_003510339.1 | 1 - 2 | 299 | 78 | |
GenBank mRNA | JI881174.1 | - | 1 - 2 | 283 | 78 | |
JP037639.1 | - | 1 - 2 | 283 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
