Assay Details
| Gene Symbol: Sdsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510810.2 | XP_003510858.1 | 8 - 9 | 966 | 66 | |
| XM_007651383.1 | XP_007649573.1 | 7 - 8 | 989 | 66 | ||
| XM_007651384.1 | XP_007649574.1 | 7 - 8 | 895 | 66 | ||
| XM_007651385.1 | XP_007649575.1 | 5 - 6 | 806 | 66 | ||
More Information
Set Membership: |
|

Sdsl
Genomic Map