Assay Details
| Gene Symbol: Gpr125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511951.2 | XP_003511999.1 | 17 - 18 | 2416 | 69 | |
| GenBank mRNA | JI885131.1 | - | 3 - 4 | 528 | 69 | |
| JP043372.1 | - | 3 - 4 | 528 | 69 | ||
More Information
Set Membership: |
|

Gpr125
Genomic Map