Assay Details
| Gene Symbol: Gpr125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511951.2 | XP_003511999.1 | 2 - 3 | 255 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr125| Gene Symbol: Gpr125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511951.2 | XP_003511999.1 | 2 - 3 | 255 | 75 | |
Set Membership: |
|