Assay Details
Gene Symbol: Ptpn20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640101.1 | XP_007638291.1 | 4 - 5 | 455 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
