Assay Details
| Gene Symbol: Akr1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497372.2 | XP_003497420.1 | 7 - 8 | 1184 | 70 | |
| GenBank mRNA | JI894009.1 | - | 7 - 8 | 1142 | 70 | |
| JP058611.1 | - | 7 - 8 | 1142 | 70 | ||
More Information
Set Membership: |
|

Akr1a1
Genomic Map