Assay Details
Gene Symbol: Tcp10l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504054.2 | XP_003504102.1 | 8 - 9 | 1065 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
