Assay Details
Gene Symbol: Rhbdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505043.2 | XP_003505091.1 | 2 - 3 | 468 | 70 | |
XM_007645550.1 | XP_007643740.1 | 2 - 3 | 468 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
