Assay Details
| Gene Symbol: Gzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509872.2 | XP_003509920.2 | 4 - 5 | 1766 | 68 | |
| XM_007650523.1 | XP_007648713.1 | 4 - 5 | 1766 | 68 | ||
| XM_007650524.1 | XP_007648714.1 | 4 - 5 | 1749 | 68 | ||
| XM_007650525.1 | XP_007648715.1 | 3 - 4 | 1650 | 68 | ||
| XM_007650526.1 | XP_007648716.1 | 4 - 5 | 1692 | 68 | ||
| GenBank mRNA | JI882005.1 | - | 2 - 3 | 225 | 68 | |
| JP062323.1 | - | 2 - 3 | 225 | 68 | ||
More Information
Set Membership: |
|

Gzf1
Genomic Map