Assay Details
| Gene Symbol: Eif1ax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499484.2 | XP_003499532.1 | 2 - 3 | 318 | 69 | |
| GenBank mRNA | JP046569.1 | - | 2 - 3 | 316 | 69 | |
More Information
Set Membership: |
|

Eif1ax
Genomic Map