Assay Details
| Gene Symbol: LOC100755061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515707.2 | XP_003515755.1 | 2 - 3 | 605 | 121 | |
| GenBank mRNA | JI866457.1 | - | 1 - 2 | 610 | 121 | |
| JP051486.1 | - | 1 - 2 | 609 | 121 | ||
More Information
Set Membership: |
|

LOC100755061
Genomic Map