Assay Details
| Gene Symbol: Sgpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499319.2 | XP_003499367.1 | 2 - 3 | 359 | 72 | |
| GenBank mRNA | JI896351.1 | - | 1 - 2 | 283 | 72 | |
More Information
Set Membership: |
|

Sgpp1
Genomic Map