Assay Details
| Gene Symbol: Ppp1r3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645603.1 | XP_007643793.1 | 1 - 2 | 81 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r3f| Gene Symbol: Ppp1r3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645603.1 | XP_007643793.1 | 1 - 2 | 81 | 62 | |
Set Membership: |
|