Assay Details
| Gene Symbol: Leprotl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499643.2 | XP_003499691.1 | 3 - 4 | 346 | 68 | |
| GenBank mRNA | JI868496.1 | - | 3 - 4 | 369 | 68 | |
| JP058545.1 | - | 3 - 4 | 369 | 68 | ||
More Information
Set Membership: |
|

Leprotl1
Genomic Map