Assay Details
Gene Symbol: Tfpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506168.2 | XP_003506216.2 | 2 - 3 | 249 | 62 | |
XM_007646624.1 | XP_007644814.1 | 4 - 5 | 442 | 62 | ||
XM_007646625.1 | XP_007644815.1 | 3 - 4 | 377 | 62 | ||
XM_007646626.1 | XP_007644816.1 | 2 - 3 | 214 | 62 | ||
XM_007646627.1 | XP_007644817.1 | 4 - 5 | 445 | 62 | ||
XM_007646628.1 | XP_007644818.1 | 2 - 3 | 239 | 62 | ||
XM_007646629.1 | XP_007644819.1 | 4 - 5 | 352 | 62 | ||
XM_007646630.1 | XP_007644820.1 | 4 - 5 | 468 | 62 | ||
XM_007646631.1 | XP_007644821.1 | 2 - 3 | 249 | 62 | ||
XM_007646632.1 | XP_007644822.1 | 2 - 3 | 249 | 62 | ||
GenBank mRNA | JP054560.1 | - | 1 - 2 | 122 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
