Assay Details
| Gene Symbol: Bscl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514302.2 | XP_003514350.1 | 3 - 4 | 891 | 57 | |
| XM_007654456.1 | XP_007652646.1 | 3 - 4 | 899 | 57 | ||
| XM_007654457.1 | XP_007652647.1 | 3 - 4 | 654 | 57 | ||
| XM_007654458.1 | XP_007652648.1 | 3 - 4 | 877 | 57 | ||
| XM_007654459.1 | XP_007652649.1 | 3 - 4 | 632 | 57 | ||
| XM_007654460.1 | XP_007652650.1 | 2 - 3 | 517 | 57 | ||
| XM_007654461.1 | XP_007652651.1 | 2 - 3 | 471 | 57 | ||
| XM_007654462.1 | XP_007652652.1 | 2 - 3 | 418 | 57 | ||
More Information
Set Membership: |
|

Bscl2
Genomic Map