Assay Details
Gene Symbol: Fam96a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640110.1 | XP_007638300.1 | 4 - 5 | 618 | 98 | |
GenBank mRNA | JI862477.1 | - | 4 - 5 | 560 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
